Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000181.3 | NP_000172.2 | 4, 5 | 858 | ||
NM_001293104.1 | NP_001280033.1 | 3, 4 | 673 | |||
NR_120531.1 | 4, 5 | 858 | ||||
XM_005250297.3 | XP_005250354.1 | 4, 5 | 853 | |||
XM_017012092.1 | XP_016867581.1 | 3, 4 | 589 | |||
GenBank mRNA | AK223406.1 | - | 4, 5 | 757 | ||
AK300223.1 | - | 4, 5 | 647 | |||
AK312752.1 | - | 4, 5 | 774 | |||
BC014142.2 | - | 4, 5 | 728 | 4662011 | ||
CD014092.1 | - | 3, 4 | 555 | |||
CD014094.1 | - | 4, 5 | 740 | |||
M15182.1 | - | 4, 5 | 753 |