Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001022.3 | NP_001013.1 | 6 | 798 | ||
NM_001321483.1 | NP_001308412.1 | 6 | 475 | |||
NM_001321484.1 | NP_001308413.1 | 6 | 605 | |||
NM_001321485.1 | NP_001308414.1 | 6 | 811 | |||
GenBank mRNA | AK311786.1 | - | 6 | 461 | ||
BC000023.2 | - | 6 | 495 | 3504083 | ||
BC007615.2 | - | 5 | 426 | 3354280 | ||
BC017386.1 | - | 6 | 464 | |||
BC018616.1 | - | 6 | 798 | 4867036 | ||
BG398397.1 | - | 6 | 605 | |||
BU603942.1 | - | 6 | 477 | |||
CB124389.1 | - | 6 | 504 | |||
CB125125.1 | - | 6 | 475 | |||
M81757.1 | - | 6 | 448 |