Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015384.4 | NP_056199.2 | 10 | 3476 | ||
NM_133433.3 | NP_597677.2 | 10 | 3476 | |||
XM_005248280.3 | XP_005248337.1 | 10 | 3465 | |||
XM_005248282.4 | XP_005248339.3 | 4 | 2967 | |||
XM_006714467.2 | XP_006714530.1 | 10 | 3465 | |||
XM_006714468.2 | XP_006714531.1 | 10 | 3465 | |||
XM_011514015.1 | XP_011512317.1 | 10 | 3465 | |||
XM_017009329.1 | XP_016864818.1 | 10 | 3465 | |||
XM_017009330.1 | XP_016864819.1 | 2 | 5739 | |||
GenBank mRNA | AJ627032.1 | - | 10 | 3445 | ||
AJ640137.1 | - | 10 | 3445 | |||
AK126804.1 | - | 6 | 2597 | |||
AK290347.1 | - | 10 | 3119 | |||
BC146821.1 | - | 10 | 3385 | |||
BX538177.1 | - | 10 | 3094 | |||
BX538178.1 | - | 10 | 3103 | |||
BX640644.1 | - | 10 | 3350 |