Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023570.3 | NP_001018864.2 | 7 | 781 | ||
NM_001023571.3 | NP_001018865.2 | 7 | 781 | |||
NM_001319107.1 | NP_001306036.1 | 7 | 780 | |||
NR_134968.1 | 7 | 780 | ||||
XM_005247911.3 | XP_005247968.1 | 7 | 780 | |||
XM_005247912.2 | XP_005247969.1 | 6 | 594 | |||
XM_011513335.2 | XP_011511637.1 | 5 | 500 | |||
XM_017007537.1 | XP_016863026.1 | 6 | 663 | |||
XM_017007538.1 | XP_016863027.1 | 3 | 465 | |||
XM_017007539.1 | XP_016863028.1 | 7 | 767 | |||
GenBank mRNA | AY964668.1 | - | 7 | 719 |