Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNG-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104124.1 | 5 | 1091 | |||
NR_104125.1 | 4 | 973 | ||||
GenBank mRNA | AK124066.1 | - | 4 | 973 |