Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LURAP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203403.1 | NP_981948.1 | 2 | 1328 | ||
GenBank mRNA | AB073606.1 | - | 2 | 1330 | ||
AK074752.1 | - | 2 | 808 | |||
AK095824.1 | - | 2 | 1327 | |||
BC036923.1 | - | 2 | 1303 | 5228515 |