Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY344640.1 | - | 1 | 1978 |
Gene Symbol: FOXD4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY344639.1 | - | 1 | 1987 |
Gene Symbol: FOXD4L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199135.4 | NP_954586.4 | 1 | 2042 | ||
GenBank mRNA | AY344641.1 | - | 1 | 2042 | ||
AY344642.1 | - | 1 | 2042 |
Gene Symbol: FOXD4L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085476.1 | NP_001078945.1 | 1 | 2549 |