Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2BFM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164416.1 | NP_001157888.1 | 3 | 811 | ||
GenBank mRNA | AK093522.1 | - | 3 | 811 |