Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2BFM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164416.1 | NP_001157888.1 | 1 | 19 | ||
XM_006724640.2 | XP_006724703.1 | 1 | 52 | |||
XM_011530922.2 | XP_011529224.1 | 1 | 215 | |||
GenBank mRNA | AK093522.1 | - | 1 | 19 |