Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AXIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004655.3 | NP_004646.3 | 2 | 692 | ||
XM_011525319.2 | XP_011523621.1 | 2 | 903 | |||
XM_011525320.1 | XP_011523622.1 | 2 | 892 | |||
XM_011525321.2 | XP_011523623.1 | 2 | 840 | |||
XM_017025192.1 | XP_016880681.1 | 2 | 559 | |||
XM_017025193.1 | XP_016880682.1 | 2 | 892 | |||
XM_017025194.1 | XP_016880683.1 | 2 | 671 | |||
GenBank mRNA | AF078165.1 | - | 1 | 492 | ||
AF205888.1 | - | 1 | 509 | |||
AK315303.1 | - | 2 | 610 | |||
BC101533.1 | - | 1 | 472 | 8069039 | ||
BC143244.1 | - | 1 | 472 | 9051745 | ||
CR933657.1 | - | 2 | 717 |