Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304548.1 | NP_001291477.1 | 32 | 5094 | ||
XM_017029452.1 | XP_016884941.1 | 32 | 5087 | |||
XM_017029453.1 | XP_016884942.1 | 32 | 5087 | |||
GenBank mRNA | AK093920.1 | - | 7 | 939 | ||
BC101698.1 | - | 6 | 793 | 8069204 | ||
BC101700.1 | - | 6 | 793 | 8069206 |