Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STON1-GTF2A1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198593.1 | NP_001185522.1 | 3, 4 | 2239 | ||
NM_001198594.1 | NP_001185523.1 | 2, 3 | 2175 | |||
NM_172311.2 | NP_758515.1 | 3, 4 | 2242 | |||
GenBank mRNA | AF026169.1 | - | 3, 4 | 2242 | ||
AK309740.1 | - | 3, 4 | 2239 | |||
BC101772.1 | - | 2, 3 | 2155 | 8069278 | ||
BC101774.1 | - | 2, 3 | 2155 | 8069280 | ||
BC143520.1 | - | 2, 3 | 2155 | 9052029 | ||
BX091338.1 | - | 2, 3 | 309 |