Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VN1R4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173857.2 | NP_776256.2 | 1 | 349 | ||
XM_011526885.1 | XP_011525187.1 | 2 | 501 | |||
XM_017026728.1 | XP_016882217.1 | 3 | 249 | |||
XM_017026729.1 | XP_016882218.1 | 3 | 267 | |||
XM_017026730.1 | XP_016882219.1 | 3 | 208 | |||
GenBank mRNA | BC104935.1 | - | 1 | 403 | 8143938 | |
BC104939.1 | - | 1 | 407 | 8143942 |