Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152467.3 | NP_689680.2 | 2 | 496 | ||
XM_011524706.2 | XP_011523008.1 | 4 | 629 | |||
GenBank mRNA | AK057224.1 | - | 2 | 459 | ||
AK301797.1 | - | 2 | 400 | |||
AK302141.1 | - | 2 | 444 | |||
AY495339.1 | - | 2 | 478 | |||
BC067753.1 | - | 2 | 483 | |||
BP371285.1 | - | 2 | 496 |