Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289396.1 | NP_001276325.1 | 30, 31 | 4577 | ||
NM_001289397.1 | NP_001276326.1 | 29, 30 | 4362 | |||
NM_001289398.1 | NP_001276327.1 | 3, 4 | 544 | |||
NM_001289399.1 | NP_001276328.1 | 4, 5 | 531 | |||
NM_001289400.1 | NP_001276329.1 | 4, 5 | 952 | |||
NM_003070.4 | NP_003061.3 | 30, 31 | 4590 | |||
NM_139045.3 | NP_620614.2 | 29, 30 | 4536 | |||
GenBank mRNA | AK094076.1 | - | 4, 5 | 952 | ||
AK298045.1 | - | 4, 5 | 531 | |||
AK300093.1 | - | 5, 6 | 596 | |||
AK308941.1 | - | 3, 4 | 689 | |||
AL558829.3 | - | 5, 6 | 536 | |||
BI758293.1 | - | 4, 5 | 599 | |||
CD107405.1 | - | 2, 3 | 299 | |||
CD513892.1 | - | 2, 3 | 340 | |||
DA084762.1 | - | 3, 4 | 541 | |||
DA823500.1 | - | 3, 4 | 467 | |||
X72889.1 | - | 30, 31 | 4578 |