Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS4L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193635.1 | NP_001180564.1 | 4, 5 | 506 | ||
NM_001193636.1 | NP_001180565.1 | 3, 4 | 749 | |||
NM_198083.3 | NP_932349.2 | 3, 4 | 548 | |||
GenBank mRNA | AK021587.1 | - | 1 | 139 | ||
BC000663.2 | - | 3, 4 | 456 | 3349226 | ||
DQ088987.1 | - | 1, 2 | 47 | |||
DQ088988.1 | - | 1, 2 | 47 | |||
GU564234.1 | - | 4, 5 | 414 | |||
GU564235.1 | - | 3, 4 | 516 |
Gene Symbol: DHRS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282988.1 | NP_001269917.1 | 2, 3 | 370 | ||
NM_001282990.1 | NP_001269919.1 | 2, 3 | 370 | |||
NM_021004.3 | NP_066284.2 | 3, 4 | 472 | |||
XM_006720005.3 | XP_006720068.1 | 3, 4 | 472 | |||
GenBank mRNA | AB045131.1 | - | 3, 4 | 497 | ||
AF044127.1 | - | 3, 4 | 443 | |||
AF064256.1 | - | 3, 4 | 446 | |||
AK001870.1 | - | 3, 4 | 438 | |||
AK314448.1 | - | 3, 4 | 437 | |||
AY358638.1 | - | 3, 4 | 453 | |||
BC003019.1 | - | 3, 4 | 420 | 2822884 | ||
BP366505.1 | - | 3, 4 | 476 | |||
DQ325464.1 | - | 2, 3 | 337 | |||
DQ338571.1 | - | 1, 2 | 199 | |||
DQ344810.1 | - | 3, 4 | 451 |