Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000350.2 | NP_000341.2 | 25 | 3778 | ||
GenBank mRNA | AB210040.1 | - | 25 | 3764 | ||
AF000148.1 | - | 25 | 3761 | |||
AF001945.1 | - | 25 | 3755 | |||
CR627391.1 | - | 3 | 190 | |||
U88667.1 | - | 25 | 3755 |