Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000033.3 | NP_000024.2 | 9 | 2314 | ||
GenBank mRNA | BC015541.1 | - | 9 | 2295 | 3896490 | |
BC025358.1 | - | 9 | 2314 | 4907640 | ||
Z21876.1 | - | 9 | 2301 |