Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000296.3 | NP_000287.3 | 34 | 10680 | ||
NM_001009944.2 | NP_001009944.2 | 34 | 10683 | |||
XM_005255370.3 | XP_005255427.1 | 24 | 8270 | |||
XM_011522528.2 | XP_011520830.1 | 35 | 10708 | |||
XM_011522529.2 | XP_011520831.1 | 35 | 10706 | |||
XM_011522530.2 | XP_011520832.1 | 35 | 10513 | |||
XM_011522532.2 | XP_011520834.1 | 34 | 10441 | |||
XM_011522534.2 | XP_011520836.1 | 35 | 10498 | |||
XM_011522535.2 | XP_011520837.1 | 26 | 8730 | |||
XM_011522537.2 | XP_011520839.1 | 24 | 7641 | |||
XM_017023285.1 | XP_016878774.1 | 25 | 8285 | |||
GenBank mRNA | AB209025.1 | - | 21 | 5037 | ||
AB209675.1 | - | 20 | 5989 | |||
L33243.1 | - | 34 | 10682 | |||
U24497.1 | - | 34 | 10609 |