Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206855.1 | NP_001193784.1 | 6 | 865 | ||
NM_003227.3 | NP_003218.2 | 9 | 1285 | |||
XM_005250553.4 | XP_005250610.1 | 10 | 1591 | |||
XM_017012573.1 | XP_016868062.1 | 10 | 1393 | |||
GenBank mRNA | AF067864.1 | - | 9 | 1271 | ||
AK000421.1 | - | 5 | 597 | |||
BC142630.1 | - | 9 | 1271 | 40147690 | ||
BC150192.1 | - | 7 | 992 | |||
BI917390.1 | - | 3 | 437 | |||
BX384750.2 | - | 6 | 866 |