Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TSC2 SIRNA Assays ›
Gene Symbol:
TSC2
Gene Name:
tuberous sclerosis 2
Gene Aliases:
LAM, PPP1R160, TSC4
Chromosome Location:
Chr.16: 2047804 - 2088720 on Build GRCh38
UniGene ID:
Hs.90303
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s502597
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TSC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000548.4
NP_000539.2
3
394
NM_001077183.2
NP_001070651.1
3
394
NM_001114382.2
NP_001107854.1
3
394
NM_001318827.1
NP_001305756.1
3
394
NM_001318829.1
NP_001305758.1
2
227
NM_001318831.1
NP_001305760.1
3
394
NM_001318832.1
NP_001305761.1
3
295
XM_005255529.4
XP_005255586.2
3
281
XM_005255531.4
XP_005255588.2
3
281
XM_011522636.2
XP_011520938.1
3
281
XM_011522637.2
XP_011520939.1
3
281
XM_011522638.2
XP_011520940.2
3
485
XM_011522639.2
XP_011520941.1
3
281
XM_011522640.2
XP_011520942.1
3
281
XM_017023615.1
XP_016879104.1
3
281
XM_017023616.1
XP_016879105.1
3
281
XM_017023617.1
XP_016879106.1
3
485
XM_017023618.1
XP_016879107.1
3
281
GenBank mRNA
AB210000.1
-
3
300
AK294548.1
-
3
292
AK295672.1
-
2
114
AK295728.1
-
3
295
AK299343.1
-
3
275
BC150300.1
-
3
269
8860381
BX647816.1
-
3
257
DA316794.1
-
3
309
DC402106.1
-
3
396
KJ534954.1
-
2
194
KJ534955.1
-
2
194
KJ534956.1
-
2
194
KJ534957.1
-
2
194
KJ534958.1
-
2
194
KJ534982.1
-
2
194
KJ534983.1
-
2
194
KJ534984.1
-
2
194
KJ534985.1
-
2
194
KJ535011.1
-
2
194
KJ535022.1
-
2
194
KJ535038.1
-
2
194
KJ535039.1
-
2
194
KJ535051.1
-
2
194
KJ535091.1
-
2
194
X75621.1
-
2
212
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
negative regulation of protein kinase activity
protein import into nucleus
endocytosis
cell cycle arrest
heart development
protein localization
negative regulation of cell proliferation
negative regulation of phosphatidylinositol 3-kinase signaling
viral process
vesicle-mediated transport
regulation of endocytosis
negative regulation of Wnt signaling pathway
negative regulation of TOR signaling
protein kinase B signaling
positive regulation of GTPase activity
regulation of insulin receptor signaling pathway
negative regulation of insulin receptor signaling pathway
insulin-like growth factor receptor signaling pathway
positive chemotaxis
regulation of small GTPase mediated signal transduction
regulation of cell cycle
negative regulation of protein kinase B signaling
GTPase activator activity
protein binding
phosphatase binding
small GTPase binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline