Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003334.3 | NP_003325.2 | 22 | 2795 | ||
NM_153280.2 | NP_695012.1 | 22 | 2723 | |||
XM_005272649.1 | XP_005272706.1 | 23 | 3173 | |||
XM_011543954.2 | XP_011542256.1 | 23 | 2815 | |||
XM_017029777.1 | XP_016885266.1 | 22 | 3890 | |||
XM_017029778.1 | XP_016885267.1 | 23 | 5064 | |||
XM_017029779.1 | XP_016885268.1 | 22 | 5572 | |||
XM_017029780.1 | XP_016885269.1 | 22 | 2680 | |||
XM_017029781.1 | XP_016885270.1 | 22 | 2728 | |||
GenBank mRNA | AF258566.1 | - | 1 | 1133 | ||
AK097343.1 | - | 6 | 1695 | |||
AK131341.1 | - | 1 | 919 | |||
AK293161.1 | - | 11 | 1387 | |||
AK296866.1 | - | 12 | 1413 | |||
BC013041.2 | - | 22 | 2662 | 3543021 | ||
BC020261.1 | - | 19 | 2357 | |||
M58028.1 | - | 22 | 2701 | |||
X52897.1 | - | 15 | 1867 | |||
X56976.1 | - | 22 | 2604 |