Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001996.3 | NP_001987.2 | 9 | 1119 | ||
NM_006485.3 | NP_006476.2 | 9 | 1119 | |||
NM_006486.2 | NP_006477.2 | 9 | 1119 | |||
NM_006487.2 | NP_006478.2 | 9 | 1119 | |||
GenBank mRNA | AF217999.1 | - | 9 | 1062 | ||
AK300806.1 | - | 9 | 1055 | |||
BC022497.1 | - | 9 | 1075 | 4819750 |