Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004643.3 | NP_004634.1 | 3 | 1763 | ||
GenBank mRNA | AK293633.1 | - | 2 | 142 | ||
BC010939.1 | - | 3 | 489 | 4333442 | ||
BI830835.1 | - | 3 | 666 | |||
BX247976.1 | - | 3 | 487 |
Gene Symbol: BCL2L2-PABPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199864.1 | NP_001186793.1 | 5 | 791 | ||
GenBank mRNA | BC011637.1 | - | 5 | 727 |