Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FARSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004461.2 | NP_004452.1 | 10 | 1087 | ||
GenBank mRNA | AF042347.1 | - | 10 | 1039 | ||
AK092093.1 | - | 10 | 1054 | |||
AK130195.1 | - | 10 | 1058 | |||
AK304587.1 | - | 9 | 966 | |||
BC006495.2 | - | 10 | 1042 | 2823551 | ||
BC035438.1 | - | 6 | 507 | |||
BC043565.1 | - | 10 | 1037 | |||
BG284015.1 | - | 1 | 20 | |||
BT007198.1 | - | 10 | 1039 | |||
CR456740.1 | - | 10 | 1039 | |||
D84471.1 | - | 10 | 1042 | |||
U07424.1 | - | 10 | 1051 |