Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144072.1 | NP_001137544.1 | 5 | 955 | ||
NM_177967.3 | NP_808882.1 | 3 | 869 | |||
NR_026644.1 | 5 | 1099 | ||||
XM_006719948.3 | XP_006720011.1 | 6 | 665 | |||
XM_011521082.2 | XP_011519384.1 | 3 | 725 | |||
XM_011521083.2 | XP_011519385.1 | 4 | 432 | |||
XM_011521084.2 | XP_011519386.1 | 5 | 520 | |||
GenBank mRNA | AK054563.1 | - | 3 | 870 | ||
AK055110.1 | - | 2 | 652 | |||
AK075419.1 | - | 5 | 526 | |||
AK075516.1 | - | 3 | 296 | |||
AK129861.1 | - | 5 | 554 | |||
AK290342.1 | - | 5 | 955 | |||
AK300908.1 | - | 5 | 441 | |||
BC063559.1 | - | 3 | 314 | |||
BC121139.1 | - | 5 | 698 | 40120987 |