Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144072.1 | NP_001137544.1 | 8 | 1344 | ||
NM_177967.3 | NP_808882.1 | 6 | 1258 | |||
NR_026644.1 | 8 | 1488 | ||||
XM_006719948.3 | XP_006720011.1 | 9 | 1054 | |||
XM_011521082.2 | XP_011519384.1 | 6 | 1114 | |||
XM_011521083.2 | XP_011519385.1 | 7 | 821 | |||
XM_011521084.2 | XP_011519386.1 | 8 | 909 | |||
XM_017020553.1 | XP_016876042.1 | 3 | 4136 | |||
GenBank mRNA | AF339812.1 | - | 3 | 328 | ||
AK054563.1 | - | 6 | 1259 | |||
AK055110.1 | - | 5 | 1041 | |||
AK075419.1 | - | 8 | 915 | |||
AK075516.1 | - | 6 | 685 | |||
AK124404.1 | - | 8 | 642 | |||
AK129861.1 | - | 8 | 943 | |||
AK290342.1 | - | 8 | 1344 | |||
AK300908.1 | - | 8 | 830 | |||
BC063559.1 | - | 6 | 703 | |||
BC072674.1 | - | 4 | 480 | |||
BC121138.1 | - | 6 | 915 | |||
BC121139.1 | - | 8 | 1087 | 40120987 |