Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004104.4 | NP_004095.4 | 42, 43 | 7523 | ||
XM_011523538.2 | XP_011521840.1 | 42, 43 | 7591 | |||
GenBank mRNA | AB209988.1 | - | 42, 43 | 7518 | ||
AY451392.1 | - | 41, 42 | 7406 | |||
BC007267.1 | - | 16, 17 | 2665 | |||
BC007909.1 | - | 11, 12 | 1938 | |||
BC014631.2 | - | 18, 19 | 3512 | |||
BC014634.2 | - | 5, 6 | 934 | |||
BC063242.1 | - | 42, 43 | 7523 | 6172538 | ||
S80437.1 | - | 7, 8 | 1277 | |||
U26644.1 | - | 41, 42 | 7388 | |||
U29344.1 | - | 42, 43 | 7523 |