Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176820.3 | NP_789790.2 | 7 | 2615 | ||
XM_011526894.2 | XP_011525196.1 | 5 | 742 | |||
XM_011526895.1 | XP_011525197.1 | 5 | 647 | |||
GenBank mRNA | AY154464.1 | - | 7 | 2587 | ||
BC136583.1 | - | 7 | 2607 | 9020573 |