Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176822.3 | NP_789792.1 | 4 | 1732 | ||
XM_011520044.1 | XP_011518346.1 | 4 | 1755 | |||
GenBank mRNA | AK310900.1 | - | 4 | 1732 | ||
AY154469.1 | - | 3 | 1409 |