Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100A7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176823.3 | NP_789793.1 | 3 | 482 | ||
GenBank mRNA | AY189118.1 | - | 3 | 482 |