Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004470.1 | NP_001004470.1 | 5 | 454 | ||
XM_017016174.1 | XP_016871663.1 | 4 | 446 | |||
GenBank mRNA | AB062101.1 | - | 5 | 454 | ||
AJ621583.1 | - | 5 | 454 | |||
BC137102.1 | - | 5 | 586 | 9021099 | ||
BC137103.1 | - | 5 | 586 | 9021100 | ||
BC144472.1 | - | 5 | 586 |