Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198489.2 | NP_940891.1 | 4 | 504 | ||
NR_104049.1 | 4 | 504 | ||||
NR_104050.1 | 4 | 504 | ||||
NR_104051.1 | 5 | 717 | ||||
XM_011542799.1 | XP_011541101.1 | 4 | 532 | |||
XM_011542800.2 | XP_011541102.1 | 2 | 212 | |||
XM_017017652.1 | XP_016873141.1 | 4 | 3286 | |||
GenBank mRNA | AB094093.1 | - | 4 | 495 | ||
BC126469.1 | - | 4 | 469 | 8992185 | ||
BC126471.1 | - | 4 | 469 | 8992187 | ||
BC144208.1 | - | 5 | 682 | |||
BC144209.1 | - | 4 | 469 | |||
BC144210.1 | - | 4 | 469 |