Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198489.2 | NP_940891.1 | 10, 11 | 1006 | ||
NR_104049.1 | 10, 11 | 1034 | ||||
NR_104050.1 | 10, 11 | 1053 | ||||
NR_104051.1 | 10, 11 | 1216 | ||||
XM_011542799.1 | XP_011541101.1 | 10, 11 | 1034 | |||
XM_011542800.2 | XP_011541102.1 | 8, 9 | 714 | |||
XM_017017652.1 | XP_016873141.1 | 10, 11 | 3788 | |||
GenBank mRNA | AB094093.1 | - | 10, 11 | 997 | ||
AW014772.1 | - | 3, 4 | 324 | |||
BC126469.1 | - | 10, 11 | 971 | 8992185 | ||
BC126471.1 | - | 10, 11 | 971 | 8992187 | ||
BC144208.1 | - | 10, 11 | 1181 | |||
BC144209.1 | - | 10, 11 | 999 | |||
BC144210.1 | - | 10, 11 | 1018 | |||
DB565963.1 | - | 3, 4 | 261 |