Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHEK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114121.2 | NP_001107593.1 | 6 | 1359 | ||
NM_001114122.2 | NP_001107594.1 | 6 | 1359 | |||
NM_001244846.1 | NP_001231775.1 | 6 | 1359 | |||
NM_001274.5 | NP_001265.2 | 6 | 702 | |||
NR_045204.1 | 5 | 1135 | ||||
NR_045205.1 | 6 | 896 | ||||
XM_011542560.2 | XP_011540862.1 | 5 | 1361 | |||
XM_011542561.2 | XP_011540863.1 | 5 | 1361 | |||
XM_011542562.2 | XP_011540864.1 | 5 | 611 | |||
XM_011542563.2 | XP_011540865.1 | 5 | 676 | |||
XM_017017146.1 | XP_016872635.1 | 6 | 546 | |||
GenBank mRNA | AB451222.1 | - | 5 | 462 | ||
AB451345.1 | - | 5 | 462 | |||
AF016582.1 | - | 5 | 496 | |||
AF032874.1 | - | 5 | 462 | |||
AK292549.1 | - | 6 | 896 | |||
AK293143.1 | - | 6 | 524 | |||
AK299783.1 | - | 5 | 296 | |||
AK300082.1 | - | 5 | 545 | |||
BC004202.2 | - | 6 | 521 | 3530606 | ||
BC017575.1 | - | 6 | 620 | 4091997 | ||
BG468734.1 | - | 5 | 561 | |||
JF289264.1 | - | 5 | 337 |