Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004019.1 | NP_001004019.1 | 6 | 1995 | ||
NM_001165035.1 | NP_001158507.1 | 6 | 2252 | |||
NM_001998.2 | NP_001989.2 | 6 | 1995 | |||
XM_006713026.2 | XP_006713089.1 | 5 | 1904 | |||
GenBank mRNA | AK129883.1 | - | 434 | |||
AK304777.1 | - | 6 | 2095 | |||
AK304827.1 | - | 6 | 2252 | |||
AY130459.1 | - | 6 | 1939 | |||
BC051690.1 | - | 6 | 1958 | 5269126 | ||
X82494.1 | - | 6 | 1939 |