Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JMJD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005920.2 | NP_001005920.2 | 4 | 431 | ||
NM_001323918.1 | NP_001310847.1 | 4 | 438 | |||
NM_001323919.1 | NP_001310848.1 | 4 | 438 | |||
NM_001323920.1 | NP_001310849.1 | 3 | 348 | |||
NM_001323922.1 | NP_001310851.1 | 3 | 348 | |||
NR_136650.1 | 4 | 438 | ||||
NR_136651.1 | 4 | 438 | ||||
NR_136652.1 | 3 | 348 | ||||
GenBank mRNA | AF370420.1 | - | 4 | 315 | ||
AL524336.3 | - | 4 | 322 | |||
AL524922.3 | - | 4 | 325 | |||
BC073785.1 | - | 3 | 403 | |||
BC094850.1 | - | 1 | 49 | |||
BC137100.1 | - | 4 | 397 | 9021097 | ||
BC137101.1 | - | 4 | 397 | 9021098 | ||
BG481039.1 | - | 4 | 334 | |||
BG826580.1 | - | 2 | 141 | |||
BI463867.1 | - | 4 | 436 | |||
BI767435.1 | - | 3 | 251 | |||
BX335015.2 | - | 4 | 318 | |||
CD674105.1 | - | 4 | 338 | |||
CN371155.1 | - | 1 | 77 | |||
DQ074695.1 | - | 3 | 221 | |||
HY023676.1 | - | 3 | 336 |