Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENPP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178543.4 | NP_848638.3 | 3 | 962 | ||
XM_011524737.1 | XP_011523039.1 | 3 | 1146 | |||
GenBank mRNA | AK126250.1 | - | 3 | 1146 | ||
AK128662.1 | - | 1 | 28 | |||
AY230663.1 | - | 3 | 945 | |||
AY358622.1 | - | 3 | 928 | |||
BC041453.1 | - | 3 | 945 | 5186743 |