Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040025.1 | NP_001035114.1 | 4, 5 | 526 | ||
GenBank mRNA | AK095847.1 | - | 2, 3 | 1654 | ||
AK304718.1 | - | 4, 5 | 365 | |||
BC042380.1 | - | 4, 5 | 364 | |||
BC063687.1 | - | 4, 5 | 328 | |||
BC105076.1 | - | 4, 5 | 526 | 8144079 | ||
BC105078.1 | - | 4, 5 | 526 | 8144081 |