Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC4G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244856.1 | NP_001231785.1 | 9 | 1234 | ||
NM_198492.3 | NP_940894.1 | 9 | 1270 | |||
GenBank mRNA | AY203939.1 | - | 9 | 1192 | ||
AY358431.1 | - | 10 | 1268 | |||
AY423706.1 | - | 9 | 1270 | |||
BC039679.1 | - | 5 | 857 |