Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC4G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244856.1 | NP_001231785.1 | 2 | 187 | ||
NM_198492.3 | NP_940894.1 | 2 | 223 | |||
GenBank mRNA | AY203939.1 | - | 2 | 145 | ||
AY358431.1 | - | 3 | 220 | |||
AY423706.1 | - | 2 | 223 | |||
BC093691.1 | - | 2 | 177 | 7939536 | ||
BC093693.1 | - | 2 | 177 | 7939538 | ||
BC143309.1 | - | 2 | 141 | 9051814 |
Gene Symbol: CLEC4GP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002931.2 | 2 | 173 | |||
GenBank mRNA | AK090689.1 | - | 2 | 173 | ||
AK090734.1 | - | 2 | 186 | |||
AK123140.1 | - | 2 | 179 |