Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RXFP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181885.2 | NP_871001.1 | 1 | 811 | ||
GenBank mRNA | AY170824.1 | - | 1 | 790 | ||
AY288415.1 | - | 1 | 790 | |||
AY394502.1 | - | 1 | 790 | |||
BC101507.1 | - | 1 | 810 | 8069013 | ||
BC101509.1 | - | 1 | 810 | 8069015 | ||
EU432129.1 | - | 1 | 790 |
Set Membership: |
G-protein coupled receptor |