Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRINP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317188.1 | NP_001304117.1 | 2 | 493 | ||
NM_199051.2 | NP_950252.1 | 2 | 697 | |||
XM_011509472.2 | XP_011507774.1 | 2 | 674 | |||
XM_011509476.2 | XP_011507778.1 | 2 | 706 | |||
XM_017001125.1 | XP_016856614.1 | 4 | 808 | |||
XM_017001126.1 | XP_016856615.1 | 3 | 673 | |||
XM_017001127.1 | XP_016856616.1 | 2 | 812 | |||
XM_017001128.1 | XP_016856617.1 | 2 | 1900 | |||
XM_017001129.1 | XP_016856618.1 | 2 | 697 | |||
XM_017001130.1 | XP_016856619.1 | 3 | 673 | |||
XM_017001131.1 | XP_016856620.1 | 2 | 717 | |||
XM_017001132.1 | XP_016856621.1 | 2 | 717 | |||
XM_017001133.1 | XP_016856622.1 | 2 | 717 | |||
GenBank mRNA | AB111893.1 | - | 2 | 444 | ||
AK123024.1 | - | 2 | 387 | |||
AK123637.1 | - | 2, 3 | 608 | |||
AK294365.1 | - | 2 | 295 | |||
BC105052.1 | - | 1 | 268 | 8144055 | ||
BC105054.1 | - | 1 | 268 | 8144057 | ||
DC330008.1 | - | 2 | 541 |