Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163857.1 | NP_001157329.1 | 3 | 567 | ||
NM_178552.3 | NP_848647.1 | 3 | 548 | |||
XM_011530165.2 | XP_011528467.1 | 4 | 780 | |||
XM_011530166.1 | XP_011528468.1 | 3 | 523 | |||
GenBank mRNA | BC042635.1 | - | 3 | 510 | 5171722 | |
CR456416.1 | - | 3 | 533 |