Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPOPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001664.2 | NP_001001664.1 | 11 | 1491 | ||
XM_005263655.4 | XP_005263712.1 | 12 | 1454 | |||
XM_005263656.4 | XP_005263713.1 | 12 | 1383 | |||
XM_005263657.4 | XP_005263714.1 | 11 | 1296 | |||
GenBank mRNA | AK293850.1 | - | 5 | 406 | ||
BC071613.1 | - | 11 | 1470 | 5275316 | ||
BX538143.1 | - | 2 | 110 |