Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESPNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194312.3 | NP_919288.2 | 4 | 813 | ||
GenBank mRNA | AK124559.1 | - | 4 | 738 |