Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024611.2 | NP_001019782.1 | 4 | 1611 | ||
XM_011534383.2 | XP_011532685.1 | 5 | 3332 | |||
XM_017008162.1 | XP_016863651.1 | 3 | 4723 | |||
GenBank mRNA | CR749811.1 | - | 4 | 1611 |