Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301738.1 | NP_001288667.1 | 5 | 653 | ||
NM_198282.3 | NP_938023.1 | 6 | 894 | |||
XM_005268445.3 | XP_005268502.1 | 6 | 824 | |||
XM_011537639.2 | XP_011535941.1 | 6 | 824 | |||
XM_011537640.2 | XP_011535942.1 | 5 | 597 | |||
GenBank mRNA | AK095896.1 | - | 4 | 1893 | ||
AK290661.1 | - | 6 | 831 | |||
AX775913.1 | - | 6 | 802 | |||
BC047779.1 | - | 6 | 811 | 5762441 | ||
FJ222241.1 | - | 4 | 560 | |||
KF430638.1 | - | 4 | 560 |