Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD34B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004441.2 | NP_001004441.2 | 5 | 822 | ||
XM_006714599.3 | XP_006714662.1 | 5 | 584 | |||
XM_011543366.2 | XP_011541668.1 | 5 | 626 | |||
XM_011543369.2 | XP_011541671.1 | 3 | 582 | |||
GenBank mRNA | BC137441.1 | - | 5 | 601 | 9021439 | |
BC137442.1 | - | 5 | 598 | 9021440 | ||
BC142957.1 | - | 4 | 502 | 8860359 | ||
CR749535.1 | - | 5 | 616 |