Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163941.1 | NP_001157413.1 | 14 | 1851 | ||
NM_001163942.1 | NP_001157414.1 | 5 | 825 | |||
NM_001163993.2 | NP_001157465.1 | 5 | 825 | |||
NM_178559.5 | NP_848654.3 | 5 | 825 | |||
XM_011515367.2 | XP_011513669.1 | 6 | 771 | |||
GenBank mRNA | AB353947.1 | - | 14 | 1851 | ||
AF399931.1 | - | 3 | 339 | |||
AY090613.1 | - | 5 | 746 | |||
AY230001.1 | - | 14 | 1744 | |||
AY234788.1 | - | 5 | 746 | |||
AY851364.1 | - | 5 | 839 | |||
AY851365.1 | - | 5 | 632 | |||
BC104894.1 | - | 5 | 575 | 8143897 | ||
BC104920.1 | - | 5 | 573 | 8143923 | ||
GU437216.1 | - | 2 | 279 | |||
GU437217.1 | - | 2 | 279 |